Anti-MRC1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 685641]

Supplier: R & D Systems
FAB25342G
102882-128EA 527.85 USD
102882-128
Anti-MRC1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 685641]
Antibodies
Type: Primary
Antigen: MRC1
Clonality: Monoclonal
Clone: 685641
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR