Anti-PVR Mouse Monoclonal Antibody (Fluorescein) [clone: 300907]

Supplier: R & D Systems
FAB25301F
220017-458EA 404.92 USD
220017-458
Anti-PVR Mouse Monoclonal Antibody (Fluorescein) [clone: 300907]
Antibodies
Type: Primary
Antigen: PVR
Clonality: Monoclonal
Clone: 300907
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR