Anti-PVR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 300907]

Supplier: R & D Systems
FAB25301A
102882-122EA 543.75 USD
102882-122
Anti-PVR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 300907]
Antibodies
Type: Primary
Antigen: PVR
Clonality: Monoclonal
Clone: 300907
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR