Anti-ADGRE5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3893]

Supplier: R & D Systems
FAB2529A
220017-456EA 519.17 USD
220017-456
Anti-ADGRE5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3893]
Antibodies
Type: Primary
Antigen: ADGRE5
Clonality: Monoclonal
Clone: 3893
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR