Anti-LIFR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 32953]

Supplier: R & D Systems
FAB249P
220017-454EA 527.85 USD
220017-454
Anti-LIFR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 32953]
Antibodies
Type: Primary
Antigen: LIFR
Clonality: Monoclonal
Clone: 32953
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR