Anti-LIFR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 32953]

Supplier: R & D Systems
FAB249A
102882-114EA 577.02 USD
102882-114
Anti-LIFR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 32953]
Antibodies
Type: Primary
Antigen: LIFR
Clonality: Monoclonal
Clone: 32953
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR