Anti-TFRC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 29806]

Supplier: R & D Systems
FAB2474P
220017-453EA 517.72 USD
220017-453
Anti-TFRC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 29806]
Antibodies
Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: 29806
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR