Anti-TGFB1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 27232]

Supplier: R & D Systems
FAB2463G
102882-110EA 561.11 USD
102882-110
Anti-TGFB1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 27232]
Antibodies
Type: Primary
Antigen: TGFB1
Clonality: Monoclonal
Clone: 27232
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR