Anti-TGFB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 27232]

Supplier: R & D Systems
FAB2463A
220017-449EA 600.15 USD
220017-449
Anti-TGFB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 27232]
Antibodies
Type: Primary
Antigen: TGFB1
Clonality: Monoclonal
Clone: 27232
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR