Anti-IFNAR1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 85228]

Supplier: R & D Systems
FAB245N
102882-108EA 543.75 USD
102882-108
Anti-IFNAR1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 85228]
Antibodies
Type: Primary
Antigen: IFNAR1
Clonality: Monoclonal
Clone: 85228
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR