Anti-IFNAR1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 85228]

Supplier: R & D Systems
FAB245C
102882-106EA 594.37 USD
102882-106
Anti-IFNAR1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 85228]
Antibodies
Type: Primary
Antigen: IFNAR1
Clonality: Monoclonal
Clone: 85228
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR