Anti-IFNAR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 85228]

Supplier: R & D Systems
FAB245A
102882-104EA 600.15 USD
102882-104
Anti-IFNAR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 85228]
Antibodies
Type: Primary
Antigen: IFNAR1
Clonality: Monoclonal
Clone: 85228
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR