Anti-ROBO4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 265703]

Supplier: R & D Systems
FAB2454P
220017-446EA 501.82 USD
220017-446
Anti-ROBO4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 265703]
Antibodies
Type: Primary
Antigen: ROBO4
Clonality: Monoclonal
Clone: 265703
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR