Anti-IL2RA Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 280406]

Supplier: R & D Systems
FAB2438P-025 FAB2438P-100
102882-098EA 339.85 USD
102882-098 102882-096
Anti-IL2RA Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 280406]
Antibodies
Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: 280406
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR