Anti-IL2RA Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 280406]

Supplier: R & D Systems
FAB2438G-100 FAB2438G-025
102882-092EA 177.88 USD
102882-092 102882-094
Anti-IL2RA Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 280406]
Antibodies
Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: 280406
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR