Anti-IL2RA Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 280406]

Supplier: R & D Systems
FAB2438A-100 FAB2438A-025
102882-086EA 485.91 USD
102882-086 102882-084
Anti-IL2RA Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 280406]
Antibodies
Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: 280406
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR