Anti-LILRB4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 293623]

Supplier: R & D Systems
FAB24251A-100 FAB24251A-025
102882-072EA 160.52 USD
102882-072 102882-074
Anti-LILRB4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 293623]
Antibodies
Type: Primary
Antigen: LILRB4
Clonality: Monoclonal
Clone: 293623
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR