Anti-TGFBR2 Mouse Monoclonal Antibody (Fluorescein) [clone: 25508]

Supplier: R & D Systems
FAB241F-100 FAB241F-025
102882-060EA 202.46 USD
102882-060 102882-062
Anti-TGFBR2 Mouse Monoclonal Antibody (Fluorescein) [clone: 25508]
Antibodies
Type: Primary
Antigen: TGFBR2
Clonality: Monoclonal
Clone: 25508
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR