Anti-TGFBR2 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 25508]

Supplier: R & D Systems
FAB241C-100 FAB241C-025
102882-056EA 316.71 USD
102882-056 102882-058
Anti-TGFBR2 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 25508]
Antibodies
Type: Primary
Antigen: TGFBR2
Clonality: Monoclonal
Clone: 25508
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR