Anti-TGFBR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 25508]

Supplier: R & D Systems
FAB241A-025 FAB241A-100
102882-054EA 614.62 USD
102882-054 102882-052
Anti-TGFBR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 25508]
Antibodies
Type: Primary
Antigen: TGFBR2
Clonality: Monoclonal
Clone: 25508
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR