Anti-TGFBR2 Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB2411P-025 FAB2411P-100
102882-048EA 261.75 USD
102882-048 102882-050
Anti-TGFBR2 Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: TGFBR2
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR