Anti-TGFBR2 Goat Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
FAB2411F-025 FAB2411F-100
102882-042EA 202.46 USD
102882-042 102882-044
Anti-TGFBR2 Goat Polyclonal Antibody (Fluorescein)
Antibodies
Type: Primary
Antigen: TGFBR2
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR