Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 296712]

Supplier: R & D Systems
FAB2410P
102882-036EA 485.91 USD
102882-036
Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 296712]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 296712
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR