Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 301040]

Supplier: R & D Systems
FAB2408P-100 FAB2408P-025
102882-032EA 501.82 USD
102882-032 102882-030
Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 301040]
Antibodies
Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: 301040
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR