Anti-NCAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 301040]

Supplier: R & D Systems
FAB2408A-100 FAB2408A-025
102882-026EA 177.88 USD
102882-026 102882-028
Anti-NCAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 301040]
Antibodies
Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: 301040
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR