Anti-CD38 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 240742]

Supplier: R & D Systems
FAB2404A
102882-022EA 454.09 USD
102882-022
Anti-CD38 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 240742]
Antibodies
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: 240742
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR