Anti-CD69 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 310106]

Supplier: R & D Systems
FAB2386P
220017-423EA 339.85 USD
220017-423
Anti-CD69 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 310106]
Antibodies
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: 310106
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR