Anti-CLEC5A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 283834]

Supplier: R & D Systems
FAB2384P
102882-020EA 527.85 USD
102882-020
Anti-CLEC5A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 283834]
Antibodies
Type: Primary
Antigen: CLEC5A
Clonality: Monoclonal
Clone: 283834
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR