Anti-CLEC5A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 283834]

Supplier: R & D Systems
FAB2384A
220017-420EA 577.02 USD
220017-420
Anti-CLEC5A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 283834]
Antibodies
Type: Primary
Antigen: CLEC5A
Clonality: Monoclonal
Clone: 283834
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR