Anti-HAVCR2 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 344823]

Supplier: R & D Systems
FAB2365N
102881-996EA 561.11 USD
102881-996
Anti-HAVCR2 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 344823]
Antibodies
Type: Primary
Antigen Symbol: HAVCR2
Antigen Name: hepatitis A virus cellular receptor 2
Antigen Synonyms: CD366, HAVcr-2, KIM-3, TIM3, TIMD-3, TIMD3, Tim-3
Clonality: Monoclonal
Clone: 344823
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR