Anti-CD69 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 298614]

Supplier: R & D Systems
FAB23591P
220017-414EA 420.83 USD
220017-414
Anti-CD69 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 298614]
Antibodies
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: 298614
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR