Anti-CCRL2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 152254]

Supplier: R & D Systems
FAB23501A
102881-990EA 577.02 USD
102881-990
Anti-CCRL2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 152254]
Antibodies
Type: Primary
Antigen: CCRL2
Clonality: Monoclonal
Clone: 152254
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR