Anti-F3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 323519]

Supplier: R & D Systems
FAB23391P
102881-988EA 513.38 USD
102881-988
Anti-F3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 323519]
Antibodies
Type: Primary
Antigen: F3
Clonality: Monoclonal
Clone: 323519
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR