Anti-LAG3 Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB2319A
102881-786EA 594.37 USD
102881-786
Anti-LAG3 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: LAG3
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR