Anti-LAG3 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 874501]

Supplier: R & D Systems
FAB23193G
102881-780EA 608.83 USD
102881-780
Anti-LAG3 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 874501]
Antibodies
Type: Primary
Antigen: LAG3
Clonality: Monoclonal
Clone: 874501
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR