Anti-LAG3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 874501]

Supplier: R & D Systems
FAB23193A
102881-778EA 647.88 USD
102881-778
Anti-LAG3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 874501]
Antibodies
Type: Primary
Antigen: LAG3
Clonality: Monoclonal
Clone: 874501
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR