Anti-IL4R Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 25463]

Supplier: R & D Systems
FAB230N-100 FAB230N-025
102881-772EA 561.11 USD
102881-772 102881-770
Anti-IL4R Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 25463]
Antibodies
Type: Primary
Antigen: IL4R
Clonality: Monoclonal
Clone: 25463
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR