Anti-IL6ST Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 28126]

Supplier: R & D Systems
FAB228P-100 FAB228P-025
102881-386EA 271.88 USD
102881-386 102881-388
Anti-IL6ST Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 28126]
Antibodies
Type: Primary
Antigen: IL6ST
Clonality: Monoclonal
Clone: 28126
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR