Anti-IL6ST Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 28126]

Supplier: R & D Systems
FAB228A-025 FAB228A-100
102881-970EA 300.8 USD
102881-970 102881-972
Anti-IL6ST Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 28126]
Antibodies
Type: Primary
Antigen: IL6ST
Clonality: Monoclonal
Clone: 28126
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR