Anti-IL6R Mouse Monoclonal Antibody (Fluorescein) [clone: 17506]

Supplier: R & D Systems
FAB227F-100 FAB227F-025
102881-962EA 193.78 USD
102881-962 102881-964
Anti-IL6R Mouse Monoclonal Antibody (Fluorescein) [clone: 17506]
Antibodies
Type: Primary
Antigen: IL6R
Clonality: Monoclonal
Clone: 17506
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR