Anti-IL6R Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 17506]

Supplier: R & D Systems
FAB227A-025 FAB227A-100
102881-960EA 577.02 USD
102881-960 102881-958
Anti-IL6R Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 17506]
Antibodies
Type: Primary
Antigen: IL6R
Clonality: Monoclonal
Clone: 17506
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR