Anti-IL17RD Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 309511]

Supplier: R & D Systems
FAB22751A
102881-946EA 552.43 USD
102881-946
Anti-IL17RD Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 309511]
Antibodies
Type: Primary
Antigen: IL17RD
Clonality: Monoclonal
Clone: 309511
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR