Anti-TNFRSF1B Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 22235]

Supplier: R & D Systems
FAB226P
220017-389EA 543.75 USD
220017-389
Anti-TNFRSF1B Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 22235]
Antibodies
Type: Primary
Antigen: TNFRSF1B
Clonality: Monoclonal
Clone: 22235
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR