Anti-TNFRSF1B Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 22235]

Supplier: R & D Systems
FAB226N
102881-944EA 519.17 USD
102881-944
Anti-TNFRSF1B Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 22235]
Antibodies
Type: Primary
Antigen: TNFRSF1B
Clonality: Monoclonal
Clone: 22235
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR