Anti-IL17RC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 309822]

Supplier: R & D Systems
FAB22691P
102881-940EA 513.38 USD
102881-940
Anti-IL17RC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 309822]
Antibodies
Type: Primary
Antigen: IL17RC
Clonality: Monoclonal
Clone: 309822
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR