Anti-TNFRSF1A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 16803]

Supplier: R & D Systems
FAB225P
220017-384EA 549.54 USD
220017-384
Anti-TNFRSF1A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 16803]
Antibodies
Type: Primary
Antigen: TNFRSF1A
Clonality: Monoclonal
Clone: 16803
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR