Anti-LDLR Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 263123]

Supplier: R & D Systems
FAB2255P
220017-381EA 517.72 USD
220017-381
Anti-LDLR Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 263123]
Antibodies
Type: Primary
Antigen: LDLR
Clonality: Monoclonal
Clone: 263123
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR