Anti-IL2RB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 27302]

Supplier: R & D Systems
FAB224P
220017-379EA 513.38 USD
220017-379
Anti-IL2RB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 27302]
Antibodies
Type: Primary
Antigen: IL2RB
Clonality: Monoclonal
Clone: 27302
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR