Anti-IL2RB Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 27302]

Supplier: R & D Systems
FAB224A
220017-377EA 577.02 USD
220017-377
Anti-IL2RB Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 27302]
Antibodies
Type: Primary
Antigen: IL2RB
Clonality: Monoclonal
Clone: 27302
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR