Anti-NCR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 253415]

Supplier: R & D Systems
FAB22491A-025 FAB22491A-100
102881-928EA 177.88 USD
102881-928 102881-930
Anti-NCR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 253415]
Antibodies
Type: Primary
Antigen: NCR2
Clonality: Monoclonal
Clone: 253415
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR